missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzophenone, 97%

Manufacturer:  Alfa Aesar™ L1146003

 View more versions of this product

Catalog No. AAL1146003



Specifications

Specifications

2-Fluoro-4-(trifluoromethyl)benzophenone
1g
C14H8F4O
8913074
VXVJVWCUWZLUIJ-UHFFFAOYSA-N
[2-fluoro-4-(trifluoromethyl)phenyl]-phenylmethanone
569577
97%
38°C to 40°C
207974-08-1
MFCD00061253
2-fluoro-4-trifluoromethyl benzophenone, 2-fluoro-4-trifluoromethyl phenyl phenyl methanone, 2-fluoro-4-trifluoromethyl phenyl phenyl methanone, 2-fluoro-4-trifluoromethyl phenyl-phenylmethanone, fxffr cf dvr, acmc-20anvd, pubchem8408, 2-fluoro-4-trifluoromethyl benzophenone, 2-fluoro-4-trifluoromethyl benzophenone, 2-fluoro-4-trifluoromethyl phenyl phenyl ketone
C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)F
268.211
268.21

Chemical Identifiers

207974-08-1
268.211
VXVJVWCUWZLUIJ-UHFFFAOYSA-N
569577
C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)C(F)(F)F)F
C14H8F4O
MFCD00061253
2-fluoro-4-trifluoromethyl benzophenone, 2-fluoro-4-trifluoromethyl phenyl phenyl methanone, 2-fluoro-4-trifluoromethyl phenyl phenyl methanone, 2-fluoro-4-trifluoromethyl phenyl-phenylmethanone, fxffr cf dvr, acmc-20anvd, pubchem8408, 2-fluoro-4-trifluoromethyl benzophenone, 2-fluoro-4-trifluoromethyl benzophenone, 2-fluoro-4-trifluoromethyl phenyl phenyl ketone
[2-fluoro-4-(trifluoromethyl)phenyl]-phenylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents