Please login to your online account to display your discounted pricing

2-Fluoro-4-(trifluoromethyl)benzoic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoic acid
CAS 115029-24-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H4F4O2
Linear Formula FC6H3(CF3)COH
Formula Weight 208.11
MDL Number MFCD00010322
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308180010

  SDS
thermo fisher scientific
308180010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystals
Formula Weight 208.11
Quantity 1g
Color White
Melting Point 165° to 169°C
CAS Max % 100.0
Linear Formula FC6H3(CF3)COH
Assay Percent Range 99%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoic acid
Molecular Formula C8H4F4O2
CAS 115029-24-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00010322
CAS Min % 98.5
Packaging Glass bottle
Synonym α,α,α,2-Tetrafluoro-p-toluic acid
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.