missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-(trifluoromethyl)benzoic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 308180010

 View more versions of this product

Catalog No. AC308180010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Fluoro-4-(trifluoromethyl)benzoic acid
165° to 169°C
Authentic
98.5
Glass bottle
≥98.5%
C8H4F4O2
208.11
α,α,α,2-Tetrafluoro-p-toluic acid
White
1g
115029-24-8
100.0
Crystals
99%
FC6H3(CF3)COH
MFCD00010322

Chemical Identifiers

115029-24-8
100.0
C8H4F4O2
208.11
α,α,α,2-Tetrafluoro-p-toluic acid
≥98.5%
98.5
99%
FC6H3(CF3)COH
MFCD00010322
2-Fluoro-4-(trifluoromethyl)benzoic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.