Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzoic acid, 98+%

$58.15 - $751.88

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoic acid
CAS 115029-24-8
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD00010322
Beilstein 8409774
Synonym alpha,alpha,alpha,2-Tetrafluoro-p-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0957103

  SDS
alfa aesar™
L0957103
1g Each for $58.15

AAL0957106

  SDS
alfa aesar™
L0957106
5g Each for $191.36

AAL0957114

  SDS
alfa aesar™
L0957114
25g Each for $751.88
Description & Specifications

Specifications

Formula Weight 208.11
Molecular Formula C8H4F4O2
MDL Number MFCD00010322
CAS 115029-24-8
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoic acid
Beilstein 8409774
Synonym alpha,alpha,alpha,2-Tetrafluoro-p-toluic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 166° to 170°C
Quantity 1g
Assay ≥98%
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.