Alfa Aesar™ 2-Fluoro-4-(trifluoromethyl)benzoic acid, 98+%
Manufacturer:  Alfa Aesar™ L0957114
 View more versions of this product
Catalog No. AAL0957114
Specifications
Specifications
2-Fluoro-4-(trifluoromethyl)benzoic acid
25g
C8H4F4O2
8409774
OCIYTBZXTFPSPI-UHFFFAOYSA-N
2-fluoro-4-(trifluoromethyl)benzoic acid
518003
≥98%
166°C to 170°C
115029-24-8
MFCD00010322
2-fluoro-4-trifluoromethyl benzoic acid, 2-fluoro-4-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-p-toluic acid, benzoic acid, 2-fluoro-4-trifluoromethyl, acmc-20aidb, pubchem2079, ksc509c3f, rarechem al bo 0632, 4-carboxy-3-fluorobenzotrifluoride, 2-fluoro-4-trifluoromethyl benzoic acid
C1=CC(=C(C=C1C(F)(F)F)F)C(=O)O
208.112
208.11
Chemical Identifiers
115029-24-8
208.112
OCIYTBZXTFPSPI-UHFFFAOYSA-N
518003
C1=CC(=C(C=C1C(F)(F)F)F)C(=O)O
C8H4F4O2
MFCD00010322
2-fluoro-4-trifluoromethyl benzoic acid, 2-fluoro-4-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-p-toluic acid, benzoic acid, 2-fluoro-4-trifluoromethyl, acmc-20aidb, pubchem2079, ksc509c3f, rarechem al bo 0632, 4-carboxy-3-fluorobenzotrifluoride, 2-fluoro-4-trifluoromethyl benzoic acid
2-fluoro-4-(trifluoromethyl)benzoic acid
Safety and Handling
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
Documents