missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-nitrotoluene, TCI America™

Manufacturer:  TCI America F02925G

 View more versions of this product

Catalog No. F02925G



Specifications

Specifications

2-Fluoro-4-nitrotoluene
5g
C7H6FNO2
2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
CC1=C(C=C(C=C1)[N+](=O)[O-])F
155.128
155.13
≥97.0% (GC)
Yellow
1427-07-2
MFCD00007199
WIQISTBTOQNVCE-UHFFFAOYSA-N
2-fluoro-1-methyl-4-nitrobenzene
74025
Crystal at 20°C

Chemical Identifiers

1427-07-2
155.128
WIQISTBTOQNVCE-UHFFFAOYSA-N
74025
CC1=C(C=C(C=C1)[N+](=O)[O-])F
C7H6FNO2
MFCD00007199
2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
2-fluoro-1-methyl-4-nitrobenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents