missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-nitrotoluene, 98%, ACROS Organics™

Chemical Identifiers

CAS 1427-07-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H6FNO2
Linear Formula CH3C6H3(NO2)F
Formula Weight 155.13
MDL Number MFCD00007199
Chemical Name or Material 2-Fluoro-4-nitrotoluene, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC207090050 SDS Thermo Fisher Scientific
207090050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1427-07-2
100.0
C7H6FNO2
155.13
2-Fluoro-4-nitrotoluene, 98%
97.5
98%
CH3C6H3(NO2)F
MFCD00007199
≥97.5%

Specifications

Yellow to brown
5g
1427-07-2
100.0
Crystalline Low Melting Solid
C7H6FNO2
155.13
05, III, 743
≥97.5%
33° to 37°C
Authentic
97.5
Glass bottle
98%
CH3C6H3(NO2)F
MFCD00007199
2-Fluoro-4-nitrotoluene, 98%

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-845-2

TSCA : TSCA