missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-nitrobenzotrifluoride, 98%

$29.62 - $346.24

Chemical Identifiers

CAS 69411-67-2
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD07782060
InChI Key XYKVQPNHGFUXBS-UHFFFAOYSA-N
Synonym 2-fluoro,4-nitrobenzotrifluoride, 2-fluoro-4-nitro-1-trifluoromethyl benzene, 2-fluoro-4-nitro-1-trifluoromethyl-benzene, 2-fluoro-4-nitrobenzotrifluoride, benzene, 2-fluoro-4-nitro-1-trifluoromethyl, pubchem1652
PubChem CID 14941411
IUPAC Name 2-fluoro-4-nitro-1-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64300MD View Documents Alfa Aesar™
H64300MD
250mg Each for $29.62
 
AAH6430003 View Documents Alfa Aesar™
H6430003
1g Each for $87.23
 
AAH6430006 View Documents Alfa Aesar™
H6430006
5g Each for $346.24
 
Specifications

Chemical Identifiers

69411-67-2
209.1
XYKVQPNHGFUXBS-UHFFFAOYSA-N
14941411
C1=CC(=C(C=C1[N+](=O)[O-])F)C(F)(F)F
C7H3F4NO2
MFCD07782060
2-fluoro,4-nitrobenzotrifluoride, 2-fluoro-4-nitro-1-trifluoromethyl benzene, 2-fluoro-4-nitro-1-trifluoromethyl-benzene, 2-fluoro-4-nitrobenzotrifluoride, benzene, 2-fluoro-4-nitro-1-trifluoromethyl, pubchem1652
2-fluoro-4-nitro-1-(trifluoromethyl)benzene

Specifications

Colorless to Yellow
69411-67-2
MFCD07782060
1881425
XYKVQPNHGFUXBS-UHFFFAOYSA-N
2-fluoro-4-nitro-1-(trifluoromethyl)benzene
14941411
Liquid
2-Fluoro-4-nitrobenzotrifluoride
250mg
C7H3F4NO2
UN2810
2-fluoro,4-nitrobenzotrifluoride, 2-fluoro-4-nitro-1-trifluoromethyl benzene, 2-fluoro-4-nitro-1-trifluoromethyl-benzene, 2-fluoro-4-nitrobenzotrifluoride, benzene, 2-fluoro-4-nitro-1-trifluoromethyl, pubchem1652
C1=CC(=C(C=C1[N+](=O)[O-])F)C(F)(F)F
209.1
209.1
98%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures