missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-nitrobenzotrifluoride, 98%

Manufacturer:  Alfa Aesar™ H6430006

 View more versions of this product

Catalog No. AAH6430006



Specifications

Specifications

2-Fluoro-4-nitrobenzotrifluoride
5g
C7H3F4NO2
UN2810
2-fluoro-4-nitro-1-trifluoromethyl benzene, 2-fluoro-4-nitrobenzotrifluoride, 2-fluoro-4-nitro-1-trifluoromethyl-benzene, benzene, 2-fluoro-4-nitro-1-trifluoromethyl, pubchem1652, 2-fluoro,4-nitrobenzotrifluoride
C1=CC(=C(C=C1[N+](=O)[O-])F)C(F)(F)F
209.1
209.1
98%
Colorless to Yellow
69411-67-2
MFCD07782060
1881425
XYKVQPNHGFUXBS-UHFFFAOYSA-N
2-fluoro-4-nitro-1-(trifluoromethyl)benzene
14941411
Liquid

Chemical Identifiers

69411-67-2
209.1
XYKVQPNHGFUXBS-UHFFFAOYSA-N
14941411
C1=CC(=C(C=C1[N+](=O)[O-])F)C(F)(F)F
C7H3F4NO2
MFCD07782060
2-fluoro-4-nitro-1-trifluoromethyl benzene, 2-fluoro-4-nitrobenzotrifluoride, 2-fluoro-4-nitro-1-trifluoromethyl-benzene, benzene, 2-fluoro-4-nitro-1-trifluoromethyl, pubchem1652, 2-fluoro,4-nitrobenzotrifluoride
2-fluoro-4-nitro-1-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

SDS Documents