missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-nitroaniline, 95%

Manufacturer:  Alfa Aesar™ H5517406

 View more versions of this product

Catalog No. AAH5517406



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C
Specifications

Specifications

2-Fluoro-4-nitroaniline
5g
C6H5FN2O2
2-fluoro-4-niroaniline
LETNCFZQCNCACQ-UHFFFAOYSA-N
2-fluoro-4-nitroaniline
101254
95%
122°C to 130°C
369-35-7
MFCD00034560
Slightly soluble in water.
C1=CC(=C(C=C1[N+](=O)[O-])F)N
156.116
156.12

Chemical Identifiers

369-35-7
156.116
LETNCFZQCNCACQ-UHFFFAOYSA-N
101254
C1=CC(=C(C=C1[N+](=O)[O-])F)N
C6H5FN2O2
MFCD00034560
2-fluoro-4-niroaniline
2-fluoro-4-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

EINECSNumber : 206-719-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents