missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-nitroaniline, 95%

Manufacturer:  Alfa Aesar™ H5517414

 View more versions of this product

Catalog No. AAH5517414



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C
Specifications

Specifications

2-Fluoro-4-nitroaniline
25g
C6H5FN2O2
2-fluoro-4-niroaniline
C1=CC(=C(C=C1[N+](=O)[O-])F)N
156.116
156.12
122°C to 130°C
369-35-7
MFCD00034560
LETNCFZQCNCACQ-UHFFFAOYSA-N
2-fluoro-4-nitroaniline
101254
95%

Chemical Identifiers

369-35-7
156.116
LETNCFZQCNCACQ-UHFFFAOYSA-N
101254
C1=CC(=C(C=C1[N+](=O)[O-])F)N
C6H5FN2O2
MFCD00034560
2-fluoro-4-niroaniline
2-fluoro-4-nitroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents