missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-methylpyridine, 98%

$44.09 - $145.42

Chemical Identifiers

CAS 461-87-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041224
InChI Key ZBFAXMKJADVOGH-UHFFFAOYSA-N
Synonym 2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, 2-fluoro-4-picoline, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j
PubChem CID 2737403
IUPAC Name 2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0160203 View Documents Alfa Aesar™
L0160203
1g Each for $44.09
 
AAL0160206 View Documents Alfa Aesar™
L0160206
5g Each for $145.42
 
Specifications

Chemical Identifiers

461-87-0
111.119
ZBFAXMKJADVOGH-UHFFFAOYSA-N
2737403
CC1=CC(=NC=C1)F
C6H6FN
MFCD00041224
2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, 2-fluoro-4-picoline, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j
2-fluoro-4-methylpyridine

Specifications

51°C (123°F)
1g
C6H6FN
UN1993
2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, 2-fluoro-4-picoline, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j
CC1=CC(=NC=C1)F
111.119
111.12
2-Fluoro-4-methylpyridine
155°C to 156°C
461-87-0
MFCD00041224
107086
ZBFAXMKJADVOGH-UHFFFAOYSA-N
2-fluoro-4-methylpyridine
2737403
98%
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.