Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-methylphenylacetic acid, 97%

$135.92 - $340.53

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-methylphenylacetic acid
CAS 518070-28-5
Molecular Formula C9H9FO2
Formula Weight 168.17
MDL Number MFCD03094329
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2664703

  SDS
alfa aesar™
H2664703
1g Each for $340.53

AAH26647MD

  SDS
alfa aesar™
H26647MD
250mg Each for $135.92
Description & Specifications

Specifications

Formula Weight 168.17
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H9FO2
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Chemical Name or Material 2-Fluoro-4-methylphenylacetic acid
MDL Number MFCD03094329
CAS 518070-28-5

Applications
As intermediates. Commonly used laboratory reagent. Used in the field of nanoparticles.

Solubility
Slightly Soluble (1.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.