missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, 97%

$310.61 - $926.57

Chemical Identifiers

CAS 874289-23-3
Molecular Formula C8H9BFNO3
Molecular Weight (g/mol) 196.972
MDL Number MFCD11856043
InChI Key STYRVOIKGOQNAD-UHFFFAOYSA-N
Synonym 2-fluoro-4-methylcarbamoyl benzeneboronic acid
PubChem CID 57497292
IUPAC Name [2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55727MD View Documents Alfa Aesar™
H55727MD
250mg Each for $310.61
 
AAH5572703 View Documents Alfa Aesar™
H5572703
1g Each for $926.57
 
Specifications

Chemical Identifiers

874289-23-3
196.972
STYRVOIKGOQNAD-UHFFFAOYSA-N
57497292
B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O
C8H9BFNO3
MFCD11856043
2-fluoro-4-methylcarbamoyl benzeneboronic acid
[2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid

Specifications

250mg
C8H9BFNO3
2-fluoro-4-methylcarbamoyl benzeneboronic acid
B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O
196.972
196.97
2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid
874289-23-3
MFCD11856043
STYRVOIKGOQNAD-UHFFFAOYSA-N
[2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
57497292
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures