missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5572703

 View more versions of this product

Catalog No. AAH5572703



Specifications

Specifications

2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid
874289-23-3
MFCD11856043
STYRVOIKGOQNAD-UHFFFAOYSA-N
[2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
57497292
97%
1g
C8H9BFNO3
2-fluoro-4-methylcarbamoyl benzeneboronic acid
B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O
196.972
196.97

Chemical Identifiers

874289-23-3
196.972
STYRVOIKGOQNAD-UHFFFAOYSA-N
57497292
B(C1=C(C=C(C=C1)C(=O)NC)F)(O)O
C8H9BFNO3
MFCD11856043
2-fluoro-4-methylcarbamoyl benzeneboronic acid
[2-fluoro-4-(methylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents