missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-methylbenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5326506

 View more versions of this product

Catalog No. AAH5326506



Specifications

Specifications

2-Fluoro-4-methylbenzeneboronic acid
5g
C7H8BFO2
2-fluoro-4-methylphenyl boronic acid, 2-fluoro-4-methylbenzeneboronic acid, 2-fluoro-4-methyl-phenyl boronic acid, boronic acid, 2-fluoro-4-methylphenyl, 2-fluoro-p-tolylboronic acid, 4-methyl-2-fluorophenylboronic acid, boronic acid, b-2-fluoro-4-methylphenyl, pubchem23775, ksc489k8t, 2-fluoro-4methylphenylboronic acid
B(C1=C(C=C(C=C1)C)F)(O)O
153.947
153.95
228°C to 233°C
170981-26-7
MFCD05664239
LIXXGOMAGHXIMP-UHFFFAOYSA-N
(2-fluoro-4-methylphenyl)boronic acid
2783132
97%

Chemical Identifiers

170981-26-7
153.947
LIXXGOMAGHXIMP-UHFFFAOYSA-N
2783132
B(C1=C(C=C(C=C1)C)F)(O)O
C7H8BFO2
MFCD05664239
2-fluoro-4-methylphenyl boronic acid, 2-fluoro-4-methylbenzeneboronic acid, 2-fluoro-4-methyl-phenyl boronic acid, boronic acid, 2-fluoro-4-methylphenyl, 2-fluoro-p-tolylboronic acid, 4-methyl-2-fluorophenylboronic acid, boronic acid, b-2-fluoro-4-methylphenyl, pubchem23775, ksc489k8t, 2-fluoro-4methylphenylboronic acid
(2-fluoro-4-methylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents