Please login to your online account to display your discounted pricing

2-Fluoro-4-methylaniline, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  320040050

 View more versions of this product

Catalog No. AC320040050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 125.16
Quantity 5g
Color Clear orange to orange-brown
Density 1.108g/mL
RTECS Number XU6650000
TSCA TSCA
CAS Max % 100.0
CAS Min % 98.5
Infrared Spectrum Authentic
Refractive Index 1.5320 to 1.5340 (20°C, 589nm)
Packaging Glass bottle
Synonym 2-Fluoro-p-toluidine
Storage Information Product darkens on storage
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Toxic if inhaled.
Health Hazard 1 GHS Signal Word: Danger
Name Note 99%
CAS 452-80-2
Linear Formula FC6H3(CH3)nH
Chemical Name or Material 2-Fluoro-4-methylaniline
MDL Number MFCD00040975
Assay Percent Range 99%
Molecular Formula C7H8FN

Specifications

Chemical Name or Material 2-Fluoro-4-methylaniline
Name Note 99%
CAS 452-80-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8FN
Linear Formula FC6H3(CH3)nH
Formula Weight 125.16
MDL Number MFCD00040975
Synonym 2-Fluoro-p-toluidine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Toxic if inhaled.

SDS & Safety Documents