missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-methylaniline, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC320040010

 View more versions of this product

Catalog No. AC320040010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Fluoro-4-methylaniline
Clear orange to orange-brown
99%
98.5
Authentic
Product darkens on storage
Liquid
99%
FC6H3(CH3)nH
MFCD00040975
1.108g/mL
1g
452-80-2
100.0
Glass bottle
1.5320 to 1.5340 (20°C, 589nm)
≥98.5%
C7H8FN
125.16
2-Fluoro-p-toluidine

Chemical Identifiers

452-80-2
100.0
C7H8FN
125.16
2-Fluoro-p-toluidine
≥98.5%
98.5
99%
FC6H3(CH3)nH
MFCD00040975
2-Fluoro-4-methylaniline, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : XU6650000

TSCA : TSCA