missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-methyl-3-nitropyridine, 98%

$28.84 - $337.73

Chemical Identifiers

CAS 19346-43-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD04112526
InChI Key AUHFWLUBEJKXLN-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-4-picoline, pyridine, 2-fluoro-4-methyl-3-nitro, 2-fluoro-3-nitro-4-methylpyridine, pubchem6297, ksc541e5r, 2-fluor-3-nitro-4-methylpyridin, 2-fluoro-4-methyl-3-nitro-pyridine, pyridine,2-fluoro-4-methyl-3-nitro, 2-fluoranyl-4-methyl-3-nitro-pyridine, 2-fluoro-3-nitro-4-picoline
PubChem CID 12408182
IUPAC Name 2-fluoro-4-methyl-3-nitropyridine
SMILES CC1=C(C(=NC=C1)F)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64837MD View Documents Alfa Aesar™
H64837MD
250mg Each for $28.84
 
AAH6483703 View Documents Alfa Aesar™
H6483703
1g Each for $85.14
 
AAH6483706 View Documents Alfa Aesar™
H6483706
5g Each for $337.73
 
Specifications

Chemical Identifiers

19346-43-1
156.116
AUHFWLUBEJKXLN-UHFFFAOYSA-N
12408182
CC1=C(C(=NC=C1)F)[N+](=O)[O-]
C6H5FN2O2
MFCD04112526
2-fluoro-3-nitro-4-picoline, pyridine, 2-fluoro-4-methyl-3-nitro, 2-fluoro-3-nitro-4-methylpyridine, pubchem6297, ksc541e5r, 2-fluor-3-nitro-4-methylpyridin, 2-fluoro-4-methyl-3-nitro-pyridine, pyridine,2-fluoro-4-methyl-3-nitro, 2-fluoranyl-4-methyl-3-nitro-pyridine, 2-fluoro-3-nitro-4-picoline
2-fluoro-4-methyl-3-nitropyridine

Specifications

33°C
19346-43-1
MFCD04112526
2-fluoro-3-nitro-4-picoline, pyridine, 2-fluoro-4-methyl-3-nitro, 2-fluoro-3-nitro-4-methylpyridine, pubchem6297, ksc541e5r, 2-fluor-3-nitro-4-methylpyridin, 2-fluoro-4-methyl-3-nitro-pyridine, pyridine,2-fluoro-4-methyl-3-nitro, 2-fluoranyl-4-methyl-3-nitro-pyridine, 2-fluoro-3-nitro-4-picoline
CC1=C(C(=NC=C1)F)[N+](=O)[O-]
156.116
156.12
98%
250mg
C6H5FN2O2
1639010
AUHFWLUBEJKXLN-UHFFFAOYSA-N
2-fluoro-4-methyl-3-nitropyridine
12408182
Solid
2-Fluoro-4-methyl-3-nitropyridine