missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-methoxyphenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC449840250

 View more versions of this product

Catalog No. AC449840250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Fluoro-4-methoxyphenylboronic acid
25g
96% min. (HPLC)
2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 2-fluoro-4-methoxyphenylboronicacid, 2-fluoro-4methoxyphenylboronic acid, 4-borono-3-fluoroanisole, acmc-209dnv, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, ksc498a9p, pubchem15367
B(C1=C(C=C(C=C1)OC)F)(O)O
169.95
169.95
Glass bottle
162101-31-7
C7H8BFO3
ULUIXJDBPYBAHS-UHFFFAOYSA-N
(2-fluoro-4-methoxyphenyl)boronic acid
15789384
97%

Chemical Identifiers

162101-31-7
169.95
2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 2-fluoro-4-methoxyphenylboronicacid, 2-fluoro-4methoxyphenylboronic acid, 4-borono-3-fluoroanisole, acmc-209dnv, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, ksc498a9p, pubchem15367
(2-fluoro-4-methoxyphenyl)boronic acid
C7H8BFO3
ULUIXJDBPYBAHS-UHFFFAOYSA-N
15789384
B(C1=C(C=C(C=C1)OC)F)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS Documents