missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-4-methoxyphenylboronic acid, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC449840050
Specifications
2-Fluoro-4-methoxyphenylboronic acid | |
96% min. (HPLC) | |
Glass bottle | |
5g | |
B(C1=C(C=C(C=C1)OC)F)(O)O | |
169.95 | |
169.95 |
162101-31-7 | |
C7H8BFO3 | |
2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 2-fluoro-4-methoxyphenylboronicacid, 2-fluoro-4methoxyphenylboronic acid, 4-borono-3-fluoroanisole, acmc-209dnv, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, ksc498a9p, pubchem15367 | |
ULUIXJDBPYBAHS-UHFFFAOYSA-N | |
(2-fluoro-4-methoxyphenyl)boronic acid | |
15789384 | |
97% |
Chemical Identifiers
162101-31-7 | |
169.95 | |
2-fluoro-4-methoxybenzeneboronic acid, 2-fluoro-4-methoxyphenyl boronic acid, 2-fluoro-4-methoxyphenylboronicacid, 2-fluoro-4methoxyphenylboronic acid, 4-borono-3-fluoroanisole, acmc-209dnv, boronic acid, 2-fluoro-4-methoxyphenyl, boronic acid, b-2-fluoro-4-methoxyphenyl, ksc498a9p, pubchem15367 | |
(2-fluoro-4-methoxyphenyl)boronic acid |
C7H8BFO3 | |
ULUIXJDBPYBAHS-UHFFFAOYSA-N | |
15789384 | |
B(C1=C(C=C(C=C1)OC)F)(O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only