Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4'-methoxybenzophenone, 99%

$54.65 - $164.70

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4'-methoxybenzophenone
CAS 66938-29-2
Molecular Formula C14H11FO2
Formula Weight 230.25
MDL Number MFCD00749331
Beilstein 2107951
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1365306

 
alfa aesar™
L1365306
5g Each for $54.65

AAL1365314

 
alfa aesar™
L1365314
25g Each for $164.70
Description & Specifications

Specifications

Formula Weight 230.25
Chemical Name or Material 2-Fluoro-4'-methoxybenzophenone
Assay 99%
Molecular Formula C14H11FO2
MDL Number MFCD00749331
CAS 66938-29-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 2107951
Health Hazard 1 Warning
Boiling Point 155° to 160°C (5mmHg)
Melting Point 55° to 57°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.