Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-methoxyaniline, JRD, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-methoxyaniline
CAS 458-52-6
Name Note JRD
Molecular Formula C7H8FNO
Formula Weight 141.15
MDL Number MFCD00047827
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3288503

  SDS
alfa aesar™
H3288503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH32885MD

  SDS
alfa aesar™
H32885MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.15
UN Number UN2811
Health Hazard 1 Danger
Chemical Name or Material 2-Fluoro-4-methoxyaniline
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Name Note JRD
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Melting Point 48° to 51°C
MDL Number MFCD00047827
CAS 458-52-6
Molecular Formula C7H8FNO
Assay 97%
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H301-H311-H373-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.