missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Fluoro-4'-methoxyacetophenone, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC263541000

 View more versions of this product

Catalog No. AC263541000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2'-Fluoro-4'-methoxyacetophenone
Authentic
80.0°C to 85.0°C (0.1mmHg)
50.0°C to 54.0°C
99%
98.5
98.5% min. (GC)
1-2-fluoro-4-methoxyphenyl ethan-1-one, 1-2-fluoro-4-methoxyphenyl ethanone, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, 2-fluoro-4-methoxyacetophenone, 4-acetyl-3-fluoroanisole, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
PIRRWUMTIBFCCW-UHFFFAOYSA-N
1-(2-fluoro-4-methoxyphenyl)ethanone
592821
Viscous Solution
>110°C
Glass bottle
Colorless
100g
74457-86-6
100.0
C9H9FO2
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
CC(=O)C1=C(C=C(C=C1)OC)F
168.17
168.17
99%

Chemical Identifiers

74457-86-6
168.17
1-2-fluoro-4-methoxyphenyl ethan-1-one, 1-2-fluoro-4-methoxyphenyl ethanone, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, 2-fluoro-4-methoxyacetophenone, 4-acetyl-3-fluoroanisole, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
1-(2-fluoro-4-methoxyphenyl)ethanone
C9H9FO2
PIRRWUMTIBFCCW-UHFFFAOYSA-N
592821
CC(=O)C1=C(C=C(C=C1)OC)F
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents