Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-methoxy-DL-phenylalanine, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-methoxy-DL-phenylalanine
Molecular Formula C10H12FNO3
Formula Weight 213.21
MDL Number MFCD09832389
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3172903

  SDS
alfa aesar™
H3172903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH31729MD

  SDS
alfa aesar™
H31729MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.21
Chemical Name or Material 2-Fluoro-4-methoxy-DL-phenylalanine
Quantity 1g
MDL Number MFCD09832389
Melting Point ≈227°C
Assay 97%
Molecular Formula C10H12FNO3