Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-iodotoluene, 99%

$40.69 - $527.36

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-iodotoluene
CAS 39998-81-7
Molecular Formula C7H6FI
Formula Weight 236.03
MDL Number MFCD00001045
Beilstein 2432998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1267906

  SDS
alfa aesar™
A1267906
5g Each for $40.69

AAA1267922

  SDS
alfa aesar™
A1267922
100g Each for $527.36
Description & Specifications

Specifications

Formula Weight 236.03
Beilstein 2432998
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 1 Warning
Chemical Name or Material 2-Fluoro-4-iodotoluene
Assay 99%
MDL Number MFCD00001045
Molecular Formula C7H6FI
CAS 39998-81-7
Boiling Point 206° to 208°C
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 22° to 24°C
Flash Point 79°C (174°F)
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.