Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-iodopyridine, 95%

$755.78 - $1,844.05

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4-iodopyridine
CAS 22282-70-8
Molecular Formula C5H3FIN
Formula Weight 222.99
MDL Number MFCD03092926
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2751703

 
alfa aesar™
H2751703
1g Each for $755.78

AAH2751706

 
alfa aesar™
H2751706
5g Each for $1,844.05
Description & Specifications

Specifications

Formula Weight 222.99
Chemical Name or Material 2-Fluoro-4-iodopyridine
Assay 95%
Molecular Formula C5H3FIN
MDL Number MFCD03092926
CAS 22282-70-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 57° to 58°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.