missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-iodo-5-methylpyridine, 95%

$43.03 - $618.39

Chemical Identifiers

CAS 153034-94-7
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD03095297
InChI Key BVKQLNXPPQEELX-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
PubChem CID 10060052
IUPAC Name 2-fluoro-4-iodo-5-methylpyridine
SMILES CC1=CN=C(C=C1I)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64983MD View Documents Alfa Aesar™
H64983MD
250mg Each for $43.03
 
AAH6498303 View Documents Alfa Aesar™
H6498303
1g Each for $127.36
 
AAH6498306 View Documents Alfa Aesar™
H6498306
5g Each for $618.39
 
Specifications

Chemical Identifiers

153034-94-7
237.016
BVKQLNXPPQEELX-UHFFFAOYSA-N
10060052
CC1=CN=C(C=C1I)F
C6H5FIN
MFCD03095297
2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
2-fluoro-4-iodo-5-methylpyridine

Specifications

Light Sensitive
40°C to 45°C
153034-94-7
MFCD03095297
2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
CC1=CN=C(C=C1I)F
237.016
237.02
95%
White
250mg
C6H5FIN
6643371
BVKQLNXPPQEELX-UHFFFAOYSA-N
2-fluoro-4-iodo-5-methylpyridine
10060052
Solid
2-Fluoro-4-iodo-5-methylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning