Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4'-hydroxybenzophenone, 98%

$45.09 - $135.31

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4'-hydroxybenzophenone
CAS 101969-75-9
Molecular Formula C13H9FO2
Formula Weight 216.21
MDL Number MFCD01075694
Synonym 4-(2-Fluorobenzoyl)phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1549403

 
alfa aesar™
L1549403
1g Each for $45.09

AAL1549406

 
alfa aesar™
L1549406
5g Each for $135.31
Description & Specifications

Specifications

Formula Weight 216.21
Chemical Name or Material 2-Fluoro-4'-hydroxybenzophenone
Assay 98%
Molecular Formula C13H9FO2
MDL Number MFCD01075694
CAS 101969-75-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Synonym 4-(2-Fluorobenzoyl)phenol
Health Hazard 1 Warning
Melting Point 115° to 118°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.