Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde, 97%

$131.15 - $523.35

Chemical Identifiers

Chemical Name or Material 2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde
Molecular Formula C9H3F7O
Formula Weight 260.11
MDL Number MFCD09258668
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3202003

  SDS
alfa aesar™
H3202003
1g Each for $131.15

AAH3202006

  SDS
alfa aesar™
H3202006
5g Each for $523.35
Description & Specifications

Specifications

Formula Weight 260.11
Chemical Name or Material 2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C9H3F7O
MDL Number MFCD09258668
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.