missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-4-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H55407MD

 View more versions of this product

Catalog No. AAH55407MD



Specifications

Specifications

2-Fluoro-4-(4-fluoro-2-nitrophenylcarbamoyl)benzeneboronic acid
1449144-94-8
MFCD20040308
JSRCTYHYPFPKJW-UHFFFAOYSA-N
[2-fluoro-4-[(4-fluoro-2-nitrophenyl)carbamoyl]phenyl]boronic acid
73996130
97%
250mg
C13H9BF2N2O5
2-fluoro-4-4-fluoro-2-nitrophenyl carbamoyl phenylboronic acid
B(C1=C(C=C(C=C1)C(=O)NC2=C(C=C(C=C2)F)[N+](=O)[O-])F)(O)O
322.031
322.03

Chemical Identifiers

1449144-94-8
322.031
JSRCTYHYPFPKJW-UHFFFAOYSA-N
73996130
B(C1=C(C=C(C=C1)C(=O)NC2=C(C=C(C=C2)F)[N+](=O)[O-])F)(O)O
C13H9BF2N2O5
MFCD20040308
2-fluoro-4-4-fluoro-2-nitrophenyl carbamoyl phenylboronic acid
[2-fluoro-4-[(4-fluoro-2-nitrophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents