Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Fluoro-3'-(trifluoromethyl)propiophenone, 97%

$214.12 - $214.12

Chemical Identifiers

Chemical Name or Material 2'-Fluoro-3'-(trifluoromethyl)propiophenone
CAS 207986-23-0
Molecular Formula C10H8F4O
Formula Weight 220.17g/mol
MDL Number MFCD00061192
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2373906

 
alfa aesar™
B23739-06
5g Each for $214.12
Description & Specifications

Specifications

Boiling Point 127° to 129°C (30mmHg)
Quantity 5g
Chemical Name or Material 2'-Fluoro-3'-(trifluoromethyl)propiophenone
CAS 207986-23-0
Assay 97%
Molecular Formula C10H8F4O
Formula Weight 220.17g/mol
MDL Number MFCD00061192
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.