missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzylamine, 97%

Manufacturer:  Alfa Aesar™ L1242803

 View more versions of this product

Catalog No. AAL1242803



Description

Description

Applications
2-Fluoro-3-(trifluoromethyl)benzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to air. Keep tightly closed. Store in a cool dry place. Store under dry inert gas. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Fluoro-3-(trifluoromethyl)benzylamine
193°C
239135-49-0
MFCD00061161
2-fluoro-3-trifluoromethyl benzylamine, 2-fluoro-3-trifluoromethylbenzylamine, 2-fluoro-3-trifluoromethyl phenyl methanamine, 2-fluoro-3-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethyl phenyl methylamine, 1-2-fluoro-3-trifluoromethyl phenyl methanamine, otf-byl-5f, acmc-209uw1, rarechem al bd 0451
C1=CC(=C(C(=C1)C(F)(F)F)F)CN
193.145
193.15
83°C (181°F)
1g
C8H7F4N
UN2735
PQUYJDPJHOSOTN-UHFFFAOYSA-N
[2-fluoro-3-(trifluoromethyl)phenyl]methanamine
2737565
97%

Chemical Identifiers

239135-49-0
193.145
PQUYJDPJHOSOTN-UHFFFAOYSA-N
2737565
C1=CC(=C(C(=C1)C(F)(F)F)F)CN
C8H7F4N
MFCD00061161
2-fluoro-3-trifluoromethyl benzylamine, 2-fluoro-3-trifluoromethylbenzylamine, 2-fluoro-3-trifluoromethyl phenyl methanamine, 2-fluoro-3-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethyl phenyl methylamine, 1-2-fluoro-3-trifluoromethyl phenyl methanamine, otf-byl-5f, acmc-209uw1, rarechem al bd 0451
[2-fluoro-3-(trifluoromethyl)phenyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents