Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzyl alcohol, 96%

$102.31 - $102.31

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzyl alcohol
CAS 207981-45-1
Molecular Formula C8H6F4O
Formula Weight 194.13
MDL Number MFCD00061166
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1999703

  SDS
alfa aesar™
A1999703
1g Each for $102.31
Description & Specifications

Specifications

Formula Weight 194.13
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzyl alcohol
Molecular Formula C8H6F4O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00061166
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point >110°C (230°F)
CAS 207981-45-1
Boiling Point 211°C
Melting Point 24° to 26°C
Health Hazard 1 Warning
Assay 96%
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.