Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoyl chloride
CAS 208173-19-7
Molecular Formula C8H3ClF4O
Formula Weight 226.56
MDL Number MFCD00061154
UN Number UN3265
Beilstein 7992377
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2346403

 
alfa aesar™
B2346403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2346406

 
alfa aesar™
B2346406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.56
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoyl chloride
Assay 97%
Molecular Formula C8H3ClF4O
MDL Number MFCD00061154
CAS 208173-19-7
UN Number UN3265
Beilstein 7992377
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluoyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 101°C (213°F)
Boiling Point 192° to 194°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.