Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzophenone, 97%

$44.15 - $111.19

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzophenone
CAS 207853-70-1
Molecular Formula C14H8F4O
Formula Weight 268.21
MDL Number MFCD00061151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1047503

 
alfa aesar™
L1047503
1g Each for $111.19

AAL10475MD

 
alfa aesar™
L10475MD
250mg Each for $44.15
Description & Specifications

Specifications

Formula Weight 268.21
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzophenone
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
CAS 207853-70-1
Molecular Formula C14H8F4O
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Health Hazard 1 Warning
Quantity 1g
Boiling Point 121°C (1mmHg)
Flash Point >110°C (230°F)
MDL Number MFCD00061151
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.