missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzophenone, 97%

Manufacturer:  Alfa Aesar™ L10475MD

 View more versions of this product

Catalog No. AAL10475MD



Specifications

Specifications

2-Fluoro-3-(trifluoromethyl)benzophenone
121°C (1mmHg)
207853-70-1
MFCD00061151
QNEOCRQLKSEYEM-UHFFFAOYSA-N
[2-fluoro-3-(trifluoromethyl)phenyl]-phenylmethanone
2737552
97%
>110°C (230°F)
250mg
C14H8F4O
2-fluoro-3-trifluoromethyl benzophenone, 2-fluoro-3-trifluoromethyl phenyl phenyl methanone, 2-fluoro-3-trifluoromethyl phenyl phenyl methanone, 2-fluoro-3-trifluoromethyl-phenyl-phenyl-methanone, 2-fluoro-3-trifluoromethyl phenyl-phenylmethanone, acmc-20anvo, pubchem8409, 2-fluoro-3-trifluoromethyl benzophenone, 2-fluoro-3-trifluoromethyl phenyl phenyl ketone, methanone, 2-fluoro-3-trifluoromethyl phenyl phenyl
C1=CC=C(C=C1)C(=O)C2=C(C(=CC=C2)C(F)(F)F)F
268.211
268.21

Chemical Identifiers

207853-70-1
268.211
QNEOCRQLKSEYEM-UHFFFAOYSA-N
2737552
C1=CC=C(C=C1)C(=O)C2=C(C(=CC=C2)C(F)(F)F)F
C14H8F4O
MFCD00061151
2-fluoro-3-trifluoromethyl benzophenone, 2-fluoro-3-trifluoromethyl phenyl phenyl methanone, 2-fluoro-3-trifluoromethyl phenyl phenyl methanone, 2-fluoro-3-trifluoromethyl-phenyl-phenyl-methanone, 2-fluoro-3-trifluoromethyl phenyl-phenylmethanone, acmc-20anvo, pubchem8409, 2-fluoro-3-trifluoromethyl benzophenone, 2-fluoro-3-trifluoromethyl phenyl phenyl ketone, methanone, 2-fluoro-3-trifluoromethyl phenyl phenyl
[2-fluoro-3-(trifluoromethyl)phenyl]-phenylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents