Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzonitrile
CAS 146070-35-1
Molecular Formula C8H3F4N
Formula Weight 189.11
MDL Number MFCD00061152
UN Number UN3276
Beilstein 8313146
Synonym 3-Cyano-2-fluorobenzotrifluoride; alpha,alpha,alpha,2-Tetrafluoro-m-tolunitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1851703

  SDS
alfa aesar™
A1851703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1851706

  SDS
alfa aesar™
A1851706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.11
Flash Point 85°C (185°F)
Assay 98%
Health Hazard 1 Danger
UN Number UN3276
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Molecular Formula C8H3F4N
MDL Number MFCD00061152
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 8313146
CAS 146070-35-1
Synonym 3-Cyano-2-fluorobenzotrifluoride; alpha,alpha,alpha,2-Tetrafluoro-m-tolunitrile
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzonitrile
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.