Please login to your online account to display your discounted pricing

2-Fluoro-3-(trifluoromethyl)benzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoic acid
CAS 115029-22-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H4F4O2
Linear Formula FC6H3(CF3)CO2H
Formula Weight 208.11
MDL Number MFCD00040980
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398190050

  SDS
thermo fisher scientific
AC398190050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398190010

  SDS
thermo fisher scientific
AC398190010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 208.11
Quantity 5g
Color White to beige
Melting Point 125° to 129°C
CAS Max % 100.0
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoic acid
Molecular Formula C8H4F4O2
Linear Formula FC6H3(CF3)CO2H
CAS 115029-22-6
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
MDL Number MFCD00040980
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.