missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzoic acid, 98%

$41.24 - $527.97

Chemical Identifiers

CAS 115029-22-6
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD00040980
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluic acid
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2028003 SDS Alfa Aesar™
B2028003
1g Each for $41.24
AAB2028014 SDS Alfa Aesar™
B2028014
25g Each for $527.97
AAB2028006 SDS Alfa Aesar™
B2028006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

115029-22-6
208.11
alpha,alpha,alpha,2-Tetrafluoro-m-toluic acid
C8H4F4O2
MFCD00040980
2-Fluoro-3-(trifluoromethyl)benzoic acid

Specifications

126° to 128°C
115029-22-6
208.11
alpha,alpha,alpha,2-Tetrafluoro-m-toluic acid
98%
1g
C8H4F4O2
MFCD00040980
2-Fluoro-3-(trifluoromethyl)benzoic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning