missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5336403

 View more versions of this product

Catalog No. AAH5336403



Description

Description

Applications
2-Fluoro-3-(trifluoromethyl)benzeneboronic acid is involved in suzuki-Miyaura coupling reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Fluoro-3-(trifluoromethyl)benzeneboronic acid
1g
C7H5BF4O2
2-fluoro-3-trifluoromethyl phenylboronic acid, 2-fluoro-3-trifluoromethyl benzeneboronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethylphenylboronic acid, 2-fluoro-3-trifluoromethyl phenylboron&, 2-fluoro-3-trifluoromethyl phenylboronicacid, boronic acid, 2-fluoro-3-trifluoromethyl phenyl, boronic acid,b-2-fluoro-3-trifluoromethyl phenyl, boronic acid, b-2-fluoro-3-trifluoromethyl phenyl
B(C1=C(C(=CC=C1)C(F)(F)F)F)(O)O
207.919
207.92
56°C to 58°C
157834-21-4
MFCD07784336
PKARAEQMGHJHMY-UHFFFAOYSA-N
[2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
16217169
97%

Chemical Identifiers

157834-21-4
207.919
PKARAEQMGHJHMY-UHFFFAOYSA-N
16217169
B(C1=C(C(=CC=C1)C(F)(F)F)F)(O)O
C7H5BF4O2
MFCD07784336
2-fluoro-3-trifluoromethyl phenylboronic acid, 2-fluoro-3-trifluoromethyl benzeneboronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethyl phenyl boronic acid, 2-fluoro-3-trifluoromethylphenylboronic acid, 2-fluoro-3-trifluoromethyl phenylboron&, 2-fluoro-3-trifluoromethyl phenylboronicacid, boronic acid, 2-fluoro-3-trifluoromethyl phenyl, boronic acid,b-2-fluoro-3-trifluoromethyl phenyl, boronic acid, b-2-fluoro-3-trifluoromethyl phenyl
[2-fluoro-3-(trifluoromethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents