Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzamide, 97%

$258.59 - $258.59

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzamide
CAS 207853-60-9
Molecular Formula C8H5F4NO
Formula Weight 207.13
MDL Number MFCD00061146
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1013506

  SDS
alfa aesar™
L1013506
5g Each for $258.59
Description & Specifications

Specifications

Formula Weight 207.13
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzamide
Assay 97%
Molecular Formula C8H5F4NO
MDL Number MFCD00061146
CAS 207853-60-9
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-toluamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 94° to 97°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.