missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzamide, 97%

Manufacturer:  Alfa Aesar™ L1013506

 View more versions of this product

Catalog No. AAL1013506



Specifications

Specifications

2-Fluoro-3-(trifluoromethyl)benzamide
5g
C8H5F4NO
2-fluoro-3-trifluoromethyl benzamide, 2-fluoro-3-trifluoromethyl-benzamide, acmc-20anvq, buttpark 45\01-73, 2-fluoranyl-3-trifluoromethyl benzamide, benzamide,2-fluoro-3-trifluoromethyl, benzamide, 2-fluoro-3-trifluoromethyl, alpha,alpha,alpha,2-tetrafluoro-m-toluamide, 2-fluoro-3-trifluoromethyl benzamide, 2-fluoro-3-trifluoromethyl-benzamide
C1=CC(=C(C(=C1)C(F)(F)F)F)C(=O)N
207.128
207.13
94°C to 97°C
207853-60-9
MFCD00061146
UHMUBYIIPNQHGD-UHFFFAOYSA-N
2-fluoro-3-(trifluoromethyl)benzamide
605688
97%

Chemical Identifiers

207853-60-9
207.128
UHMUBYIIPNQHGD-UHFFFAOYSA-N
605688
C1=CC(=C(C(=C1)C(F)(F)F)F)C(=O)N
C8H5F4NO
MFCD00061146
2-fluoro-3-trifluoromethyl benzamide, 2-fluoro-3-trifluoromethyl-benzamide, acmc-20anvq, buttpark 45\01-73, 2-fluoranyl-3-trifluoromethyl benzamide, benzamide,2-fluoro-3-trifluoromethyl, benzamide, 2-fluoro-3-trifluoromethyl, alpha,alpha,alpha,2-tetrafluoro-m-toluamide, 2-fluoro-3-trifluoromethyl benzamide, 2-fluoro-3-trifluoromethyl-benzamide
2-fluoro-3-(trifluoromethyl)benzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents