Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%

$166.24 - $166.24

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzaldehyde
CAS 112641-20-0
Molecular Formula C8H4F4O
Formula Weight 192.11
MDL Number MFCD00061246
Beilstein 8311752
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-tolualdehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2373806

  SDS
alfa aesar™
B2373806
5g Each for $166.24
Description & Specifications

Specifications

Formula Weight 192.11
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C8H4F4O
MDL Number MFCD00061246
CAS 112641-20-0
Beilstein 8311752
Synonym alpha,alpha,alpha,2-Tetrafluoro-m-tolualdehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 73°C (163°F)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.