Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(trifluoromethyl)aniline, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)aniline
CAS 123973-25-1
Molecular Formula C7H5F4N
Formula Weight 179.12
MDL Number MFCD00061241
UN Number UN2810
Synonym 3-Amino-2-fluorobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1690603

  SDS
alfa aesar™
A1690603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1690606

  SDS
alfa aesar™
A1690606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.12
Molecular Formula C7H5F4N
CAS 123973-25-1
Assay 97%
Flash Point 82°C (179°F)
Synonym 3-Amino-2-fluorobenzotrifluoride
MDL Number MFCD00061241
Health Hazard 2 GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Danger
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)aniline
Boiling Point 189°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 1g
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN2810
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.