Learn More
2-Fluoro-3-nitropyridine, 98%
CAS: 1480-87-1 | C5H3FN2O2 | 142.09 g/mol
Supplier: Thermo Scientific Chemicals H6126022
Description
2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoro-3-nitropyridine is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 2-Fluoro-3-nitropyridine | |
| 18°C | |
| 110°C (10 mmHg) | |
| MFCD03095068 | |
| 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n | |
| QDKIYDGHCFZBGC-UHFFFAOYSA-N | |
| 2-fluoro-3-nitropyridine | |
| 2762817 | |
| 98% |
| 1480-87-1 | |
| Yellow | |
| C5H3FN2O2 | |
| 100 g | |
| Slightly soluble in water. | |
| [O-][N+](=O)C1=C(F)N=CC=C1 | |
| 142.09 | |
| 142.09 | |
| Low Melting Solid |
Chemical Identifiers
| 1480-87-1 | |
| 142.09 | |
| QDKIYDGHCFZBGC-UHFFFAOYSA-N | |
| 2762817 | |
| [O-][N+](=O)C1=C(F)N=CC=C1 |
| C5H3FN2O2 | |
| MFCD03095068 | |
| 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n | |
| 2-fluoro-3-nitropyridine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only