missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-nitrobenzotrifluoride, 98%

$31.30 - $365.49

Chemical Identifiers

CAS 61324-97-8
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD08532487
InChI Key PGKWPKDZNHZBQK-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitrobenzotrifluoride, 2-fluoro-1-nitro-3-trifluoromethyl benzene, benzene, 2-fluoro-1-nitro-3-trifluoromethyl, 2-fluoro-3-trifluoromethyl nitrobenzene, 2-fluoro-3-trifluoromethylnitrobenzene, 2-fluoro-3-nitrobenzotrifluori
PubChem CID 23340645
IUPAC Name 2-fluoro-1-nitro-3-(trifluoromethyl)benzene
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])F)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64562MD View Documents Alfa Aesar™
H64562MD
250mg Each for $31.30
 
AAH6456203 View Documents Alfa Aesar™
H6456203
1g Each for $91.91
 
AAH6456206 View Documents Alfa Aesar™
H6456206
5g Each for $365.49
 
Specifications

Chemical Identifiers

61324-97-8
209.1
PGKWPKDZNHZBQK-UHFFFAOYSA-N
23340645
C1=CC(=C(C(=C1)[N+](=O)[O-])F)C(F)(F)F
C7H3F4NO2
MFCD08532487
2-fluoro-3-nitrobenzotrifluoride, 2-fluoro-1-nitro-3-trifluoromethyl benzene, benzene, 2-fluoro-1-nitro-3-trifluoromethyl, 2-fluoro-3-trifluoromethyl nitrobenzene, 2-fluoro-3-trifluoromethylnitrobenzene, 2-fluoro-3-nitrobenzotrifluori
2-fluoro-1-nitro-3-(trifluoromethyl)benzene

Specifications

250mg
C7H3F4NO2
UN2811
2-fluoro-3-nitrobenzotrifluoride, 2-fluoro-1-nitro-3-trifluoromethyl benzene, benzene, 2-fluoro-1-nitro-3-trifluoromethyl, 2-fluoro-3-trifluoromethyl nitrobenzene, 2-fluoro-3-trifluoromethylnitrobenzene, 2-fluoro-3-nitrobenzotrifluori
C1=CC(=C(C(=C1)[N+](=O)[O-])F)C(F)(F)F
209.1
209.1
98%
61324-97-8
MFCD08532487
13320950
PGKWPKDZNHZBQK-UHFFFAOYSA-N
2-fluoro-1-nitro-3-(trifluoromethyl)benzene
23340645
Solid
2-Fluoro-3-nitrobenzotrifluoride
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.