missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-nitro-6-methylpyridine, 98%

Manufacturer:  Alfa Aesar™ H64103MD

 View more versions of this product

Catalog No. AAH64103MD



Description

Description

Applications
6-Methyl-3-nitro-2-(2-phenylhydrazinyl)pyridine (MNPHP) was synthesized from 2-fluoro-3-nitro-6-methylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Fluoro-3-nitro-6-methylpyridine
30°C
19346-45-3
MFCD03095067
2-fluoro-3-nitro-6-picoline, 2-fluoro-3-nitro-6-methylpyridine, 2-fluoro-6-methyl-3-nitro-pyridine, 2-fluoro-6-methyl-3-nitro pyridine, pyridine, 2-fluoro-6-methyl-3-nitro, pubchem6309, abbypharma ap-30-0928, 6-fluoro-5-nitro-2-picoline, 2-fluoranyl-6-methyl-3-nitro-pyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])F
156.116
156.12
98%
White
250mg
C6H5FN2O2
1638807
BCCWOQXQVNCXKC-UHFFFAOYSA-N
2-fluoro-6-methyl-3-nitropyridine
2783178
Solid

Chemical Identifiers

19346-45-3
156.116
BCCWOQXQVNCXKC-UHFFFAOYSA-N
2783178
CC1=NC(=C(C=C1)[N+](=O)[O-])F
C6H5FN2O2
MFCD03095067
2-fluoro-3-nitro-6-picoline, 2-fluoro-3-nitro-6-methylpyridine, 2-fluoro-6-methyl-3-nitro-pyridine, 2-fluoro-6-methyl-3-nitro pyridine, pyridine, 2-fluoro-6-methyl-3-nitro, pubchem6309, abbypharma ap-30-0928, 6-fluoro-5-nitro-2-picoline, 2-fluoranyl-6-methyl-3-nitro-pyridine
2-fluoro-6-methyl-3-nitropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents