Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-methylpyridine, 97%

$56.04 - $207.80

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-methylpyridine
CAS 2369-18-8
Molecular Formula C6H6FN
Formula Weight 111.12
MDL Number MFCD00041223
Beilstein 1877
Synonym 2-Fluoro-3-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1070103

  SDS
alfa aesar™
L1070103
1g Each for $56.04

AAL1070106

  SDS
alfa aesar™
L1070106
5g Each for $207.80
Description & Specifications

Specifications

Formula Weight 111.12
Chemical Name or Material 2-Fluoro-3-methylpyridine
Assay 97%
Molecular Formula C6H6FN
MDL Number MFCD00041223
CAS 2369-18-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 1877
Synonym 2-Fluoro-3-picoline
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >100°C (212°F)
Boiling Point 148° to 150°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.