missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-methylpyridine-5-boronic acid, 97%

$36.39 - $428.70

Chemical Identifiers

CAS 904326-92-7
Molecular Formula C6H7BFNO2
Molecular Weight (g/mol) 154.935
MDL Number MFCD03411562
InChI Key XCWIMBMPJGQNHG-UHFFFAOYSA-N
Synonym 2-fluoro-3-methylpyridine-5-boronic acid, 6-fluoro-5-methylpyridin-3-yl boronic acid, 2-fluoro-3-methylpyridine-5-yl boronic acid, 2-fluoro-3-methyl-5-pyridineboronic acid, 6-fluoro-5-methylpyridine-3-boronic acid, 6-fluoro-5-methylpyridin-3-yl-3-boronic acid, 2-fluoro-3-picoline-5-boronic acid, 6-fluoro-5-methylpyridine-3-boronicacid, 6-fluoro-5-methyl-3-pyridineboronic acid, 6-fluoro-5-methyl-3-pyridyl boronic acid
PubChem CID 17750247
IUPAC Name (6-fluoro-5-methylpyridin-3-yl)boronic acid
SMILES B(C1=CC(=C(N=C1)F)C)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64157MD View Documents Alfa Aesar™
H64157MD
250mg Each for $36.39
 
AAH6415703 View Documents Alfa Aesar™
H6415703
1g Each for $107.64
 
AAH6415706 View Documents Alfa Aesar™
H6415706
5g Each for $428.70
 
Specifications

Chemical Identifiers

904326-92-7
154.935
XCWIMBMPJGQNHG-UHFFFAOYSA-N
17750247
B(C1=CC(=C(N=C1)F)C)(O)O
C6H7BFNO2
MFCD03411562
2-fluoro-3-methylpyridine-5-boronic acid, 6-fluoro-5-methylpyridin-3-yl boronic acid, 2-fluoro-3-methylpyridine-5-yl boronic acid, 2-fluoro-3-methyl-5-pyridineboronic acid, 6-fluoro-5-methylpyridine-3-boronic acid, 6-fluoro-5-methylpyridin-3-yl-3-boronic acid, 2-fluoro-3-picoline-5-boronic acid, 6-fluoro-5-methylpyridine-3-boronicacid, 6-fluoro-5-methyl-3-pyridineboronic acid, 6-fluoro-5-methyl-3-pyridyl boronic acid
(6-fluoro-5-methylpyridin-3-yl)boronic acid

Specifications

250mg
C6H7BFNO2
2-fluoro-3-methylpyridine-5-boronic acid, 6-fluoro-5-methylpyridin-3-yl boronic acid, 2-fluoro-3-methylpyridine-5-yl boronic acid, 2-fluoro-3-methyl-5-pyridineboronic acid, 6-fluoro-5-methylpyridine-3-boronic acid, 6-fluoro-5-methylpyridin-3-yl-3-boronic acid, 2-fluoro-3-picoline-5-boronic acid, 6-fluoro-5-methylpyridine-3-boronicacid, 6-fluoro-5-methyl-3-pyridineboronic acid, 6-fluoro-5-methyl-3-pyridyl boronic acid
B(C1=CC(=C(N=C1)F)C)(O)O
154.935
154.94
97%
904326-92-7
MFCD03411562
XCWIMBMPJGQNHG-UHFFFAOYSA-N
(6-fluoro-5-methylpyridin-3-yl)boronic acid
17750247
Powder
2-Fluoro-3-methylpyridine-5-boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning